Learn More
Medchemexpress LLC 2,3-Dihydroxy-4-methoxyacetophenone | 708-53-2 | 100 MG
SDP
Catalog No. 5000418277
Encompass_Preferred
2,3-Dihydroxy-4-methoxyacetophenone, derived from *Cynanchum paniculatum*, is a neuroprotective compound with potential for Alzheimer's disease research. It improves cognitive function and protects HT22 cells from glutamate-induced cell death by inhibiting Ca2+ accumulation and exhibiting antioxidative activity. *In vivo*, it has shown to improve scopolamine-induced spatial memory impairment.
  • Derived from *Cynanchum paniculatum*
  • Exhibits neuroprotective activity
  • Protects HT22 cells from glutamate-induced cell death
  • Inhibits Ca2+ accumulation
  • Demonstrates antioxidative activity
  • Improves scopolamine-induced spatial memory impairment *in vivo*
Catalog No. 50-004-18277 Supplier Medchemexpress LLC Supplier No. HYN7509100MG
2,3-Dihydroxy-4-methoxyacetophenone, derived from *Cynanchum paniculatum*, is a neuroprotective compound with potential for Alzheimer's disease research. It improves cognitive function and protects HT22 cells from glutamate-induced cell death by inhibiting Ca2+ accumulation and exhibiting antioxidative activity. *In vivo*, it has shown to improve scopolamine-induced spatial memory impairment.
  • Derived from *Cynanchum paniculatum*
  • Exhibits neuroprotective activity
  • Protects HT22 cells from glutamate-induced cell death
  • Inhibits Ca2+ accumulation
  • Demonstrates antioxidative activity
  • Improves scopolamine-induced spatial memory impairment *in vivo*