Learn More
Medchemexpress LLC 2,3-Dihydroxy-4-methoxyacetophenone | 708-53-2 | 50 MG
SDP
Catalog No. 5000417291
Encompass_Preferred
2,3-Dihydroxy-4-methoxyacetophenone is a neuroprotective compound isolated from *Cynanchum paniculatum*. It protects HT22 cells from glutamate-induced cell death, inhibits [Ca2+] accumulation, and exhibits antioxidative activity. This compound improves cognitive function and shows potential for research in Alzheimer's disease.
  • Neuroprotective compound derived from *Cynanchum paniculatum*.
  • Improves cognitive function.
  • Protects HT22 cells from glutamate-induced cell death.
  • Inhibits [Ca2+] accumulation in HT22 cells.
  • Exhibits antioxidative activity.
  • Improves impairment of spatial memory induced by scopolamine.
  • Appears as a white to yellow solid.
Catalog No. 50-004-17291 Supplier Medchemexpress LLC Supplier No. HYN750950MG
2,3-Dihydroxy-4-methoxyacetophenone is a neuroprotective compound isolated from *Cynanchum paniculatum*. It protects HT22 cells from glutamate-induced cell death, inhibits [Ca2+] accumulation, and exhibits antioxidative activity. This compound improves cognitive function and shows potential for research in Alzheimer's disease.
  • Neuroprotective compound derived from *Cynanchum paniculatum*.
  • Improves cognitive function.
  • Protects HT22 cells from glutamate-induced cell death.
  • Inhibits [Ca2+] accumulation in HT22 cells.
  • Exhibits antioxidative activity.
  • Improves impairment of spatial memory induced by scopolamine.
  • Appears as a white to yellow solid.