Learn More
Selleck Chemical LLC 2.3-Dihydroxybenzoic acid S4946-25mg
SDP
Catalog No. 503108843
Encompass_Preferred
2 3-Dihydroxybenzoic acid (Pyrocatechuic acid 3-Hydroxysalicylic acid Hypogallic acid) is a natural phenol found in Phyllanthus acidus and in the aquatic fern Salvinia molesta also a product of human aspirin metabolism It is a potentially useful iron-chelating drug and has antimicrobial properties
Catalog No. 50-310-8843 Supplier Selleck Chemical LLC Supplier No. S494625MG
2 3-Dihydroxybenzoic acid (Pyrocatechuic acid 3-Hydroxysalicylic acid Hypogallic acid) is a natural phenol found in Phyllanthus acidus and in the aquatic fern Salvinia molesta also a product of human aspirin metabolism It is a potentially useful iron-chelating drug and has antimicrobial properties