Learn More
Medchemexpress LLC 2,3-dihydroxybenzoic acid | 303-38-8 | MFCD00002446 | 99.6% | 154.12 | C7H6O4 | 250mg
SDP
Catalog No. 5000268026
Encompass_Preferred
Pyrocatechuic acid (Standard) is the analytical standard of Pyrocatechuic acid This product is intended for research and analytical applications Pyrocatechuic acid is a normal human benzoic acid metabolite found in plasma and has increased levels after aspirin ingestion
Catalog No. 50-002-68026 Supplier Medchemexpress LLC Supplier No. HYY0202R250MG
Pyrocatechuic acid (Standard) is the analytical standard of Pyrocatechuic acid This product is intended for research and analytical applications Pyrocatechuic acid is a normal human benzoic acid metabolite found in plasma and has increased levels after aspirin ingestion