Learn More
Medchemexpress LLC 2,3-Dihydroxynaphthalene | 92-44-4 | 98.6% | 160.17 | 250 G
SDP
Catalog No. 5000451333
Encompass_Preferred
Naphthalene-2,3-diol is a biochemical reagent used as a biological material or organic compound for life science related research. It is utilized in various applications, including the production of dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.
  • Can be used as a biological material
  • Can be used as an organic compound for life science research
  • Used in making dyes and pigments
  • Used for fluorescent whiteners and tanning agents
  • Acts as an antioxidant and antiseptic
Catalog No. 50-004-51333 Supplier Medchemexpress LLC Supplier No. 250GHYW012558
Naphthalene-2,3-diol is a biochemical reagent used as a biological material or organic compound for life science related research. It is utilized in various applications, including the production of dyes, pigments, fluorescent whiteners, tanning agents, antioxidants, and antiseptics.
  • Can be used as a biological material
  • Can be used as an organic compound for life science research
  • Used in making dyes and pigments
  • Used for fluorescent whiteners and tanning agents
  • Acts as an antioxidant and antiseptic