Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydroxypyridine, 98%, Thermo Scientific™

Catalog No. AC153560250
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153560250 25 g
AC153561000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153560250 Supplier Thermo Scientific Chemicals Supplier No. 153560250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16867-04-2
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00006271
InChI Key GGOZGYRTNQBSSA-UHFFFAOYSA-N
Synonym 2,3-dihydroxypyridine, 2,3-pyridinediol, pyridine-2,3-diol, 3-hydroxy-2-pyridone, 2 1h-pyridinone, 3-hydroxy, 3-hydroxy-2 1h-pyridinone, 3-hydroxypyridin-2 1h-one, 2 1h-pyridone, 3-hydroxy, hydroxypyridone, unii-7hy4bcj130
PubChem CID 28115
IUPAC Name 3-hydroxy-1H-pyridin-2-one
SMILES C1=CNC(=O)C(=C1)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown or Gray to Khaki
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula (C5H3N)(OH)2
Quantity 25 g
Beilstein 21, II, 107
Formula Weight 111.1
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dihydroxypyridine, 98%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.