Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimercapto-1-propanol, Solstice

Catalog No. 6000111 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-001-11 5 mL
60-001-10 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-001-11 Supplier Solstice Supplier No. 385205ML

missing translation for 'validMainframeMsg'

for complexometry, for spectrophotometric det. of Cu,≥98.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.216
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

Density 1.239g/mL at 25°C
Flash Point 112°C (233.6°F)
Packaging Glass Bottle
Linear Formula HOCH2CH(SH)CH2SH
Quantity 5 mL
UN Number UN2810
Beilstein 1732058
Formula Weight 124.23g/mol
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimercapto-1-propanol
Hazard Statement P261-P301 + P310-P305 + P351 + P338
RTECSNumber UB2625000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.