Learn More
Medchemexpress LLC 2,3-dimethoxy-1,4-naphthoquinone | 6956-96-3 | 99.8% | 10 MG
SDP
Catalog No. 5000233863
Encompass_Preferred
DMNQ is a redox-cycling small molecule that produces hydrogen peroxide and increases intracellular reactive oxygen species (ROS). It is supplied as a solid powder for research use and is commonly used to model oxidative stress in cell-based assays.
  • Redox-cycling agent that induces ROS production.
  • Solid powder with formula C12H10O4 and molecular weight 218.21 g/mol.
  • High purity suitable for research applications.
  • Available in small milligram package sizes for laboratory use.
  • Store powder at -20°C; store solutions at -80°C for long-term storage.
Catalog No. 50-002-33863 Supplier Medchemexpress LLC Supplier No. HY12102610MG
DMNQ is a redox-cycling small molecule that produces hydrogen peroxide and increases intracellular reactive oxygen species (ROS). It is supplied as a solid powder for research use and is commonly used to model oxidative stress in cell-based assays.
  • Redox-cycling agent that induces ROS production.
  • Solid powder with formula C12H10O4 and molecular weight 218.21 g/mol.
  • High purity suitable for research applications.
  • Available in small milligram package sizes for laboratory use.
  • Store powder at -20°C; store solutions at -80°C for long-term storage.