Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-dimethoxy-13-(2-(pyrrolidin-1-yl)ethoxy)-dioxolo[4',5':4,5]benzo[1,2-c]phenanthridine | 2248702-80-7 | 98.3% | 5 MG
SDP

Catalog No. 5000233901
Encompass_Preferred

TDP1 Inhibitor-1 is a small-molecule research compound that inhibits Tyrosyl-DNA phosphodiesterase 1 (TDP1) and is used to probe DNA repair and cytotoxicity in cell-based cancer research.

  • Inhibits TDP1; reported IC50 ≈ 7 μM.
  • Molecular weight 446.5 g/mol; formula C26H26N2O5.
  • Purity 98.3% (reported by supplier).
  • Supplied in small milligram pack sizes suitable for laboratory studies.
  • Intended for research use only; not for human or clinical use.

Catalog No. 50-002-33901 Supplier Medchemexpress LLC Supplier No. HY1193725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TDP1 Inhibitor-1 is a small-molecule research compound that inhibits Tyrosyl-DNA phosphodiesterase 1 (TDP1) and is used to probe DNA repair and cytotoxicity in cell-based cancer research.

  • Inhibits TDP1; reported IC50 ≈ 7 μM.
  • Molecular weight 446.5 g/mol; formula C26H26N2O5.
  • Purity 98.3% (reported by supplier).
  • Supplied in small milligram pack sizes suitable for laboratory studies.
  • Intended for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.