Learn More
Medchemexpress LLC 2,3-Dimethoxybenzaldehyde | 86-51-1 | 99.9% | 166.18 | 500 G
SDP
Catalog No. 5000404918
Encompass_Preferred
2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog with high antifungal activity (MIC=2.5 mM). It can be used for the synthesis of berberine. It also exhibits weak antiviral activity against HSV-1 replication (EC50 of 230.9 μM) and antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • Benzaldehyde analog with high antifungal activity.
  • Can be used for the synthesis of berberine.
  • Exhibits weak antiviral activity against HSV-1 replication.
  • Exhibits antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • Solid appearance.
  • Off-white to light yellow color.
  • Soluble in DMSO.
Catalog No. 50-004-04918 Supplier Medchemexpress LLC Supplier No. HY41407500G
2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog with high antifungal activity (MIC=2.5 mM). It can be used for the synthesis of berberine. It also exhibits weak antiviral activity against HSV-1 replication (EC50 of 230.9 μM) and antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • Benzaldehyde analog with high antifungal activity.
  • Can be used for the synthesis of berberine.
  • Exhibits weak antiviral activity against HSV-1 replication.
  • Exhibits antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • Solid appearance.
  • Off-white to light yellow color.
  • Soluble in DMSO.