Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxybenzaldehyde, 98+%

Catalog No. AAA1408022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1408022 100 g
AAA1408014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1408022 Supplier Thermo Scientific Chemicals Supplier No. A1408022
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 86-51-1 | C9H10O3 | 166.176 g/mol

2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxybenzaldehyde possess antifungal activity of redox-active benzaldehydes that target cellular antioxidation. It could function as chemosensitizing agents in concert with conventional drugs to improve antifungal efficacy.

Solubility
Solubility in methanol with almost transparency.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-51-1
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00003309
InChI Key JIVGSHFYXPRRSZ-UHFFFAOYSA-N
Synonym o-veratraldehyde, benzaldehyde, 2,3-dimethoxy, unii-8alp3sy00l, 2,3-dimethoxy-benzaldehyde, 8alp3sy00l, benzaldehyde,3-dimethoxy, dimethoxybenzaldehyde, pubchem8216, benzaldehyde, dimethoxy, upcmld00wstructure66
PubChem CID 66581
IUPAC Name 2,3-dimethoxybenzaldehyde
SMILES COC1=CC=CC(=C1OC)C=O

Specifications

Melting Point 51°C to 52°C
Boiling Point 178°C (20 mmHg)
Flash Point >110°C (230°F)
Quantity 100 g
Beilstein 908264
Sensitivity Air Sensitive
Formula Weight 166.18
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-677-7
RTECSNumber CU5732000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.