Learn More
Medchemexpress LLC 2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) | 86-51-1 | 100.0% | 166.18 | 100 G
SDP
Catalog No. 5000404606
Encompass_Preferred
2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog. It possesses high antifungal activity, with a Minimum Inhibitory Concentration (MIC) of 2.5 mM. This compound can also be utilized in the synthesis of berberine. Additionally, it shows weak antiviral activity against HSV-1 replication and antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • It is a benzaldehyde analog.
  • It shows high antifungal activity.
  • It can be used for the synthesis of berberine.
  • It exhibits weak antiviral activity.
  • It has antibacterial activity.
Catalog No. 50-004-04606 Supplier Medchemexpress LLC Supplier No. HY41407100G
2,3-Dimethoxybenzaldehyde (o-Veratraldehyde) is a benzaldehyde analog. It possesses high antifungal activity, with a Minimum Inhibitory Concentration (MIC) of 2.5 mM. This compound can also be utilized in the synthesis of berberine. Additionally, it shows weak antiviral activity against HSV-1 replication and antibacterial activity against E. coli BL-21 and S. aureus NR-46003.
  • It is a benzaldehyde analog.
  • It shows high antifungal activity.
  • It can be used for the synthesis of berberine.
  • It exhibits weak antiviral activity.
  • It has antibacterial activity.