Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxycinnamic acid, predominantly trans, 98+%

Catalog No. AAA1593630
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1593630 250 g
AAA1593609 10 g
AAA1593618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1593630 Supplier Thermo Scientific Chemicals Supplier No. A1593630
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7345-82-6 | C11H12O4 | 208.213 g/mol

2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2E,2E)-N,N,N-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3-Dimethoxycinnamic acid is used in the synthesis of (2E,2′E,2′′E)-N,N′,N′′-(nitrilotri-2,1-ethanediyl)tris[3-(2,3-dimethoxyphenyl)-2-propenamide].

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7345-82-6
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004376
InChI Key QAXPUWGAGVERSJ-VOTSOKGWSA-N
Synonym 2,3-dimethoxycinnamic acid, trans-2,3-dimethoxycinnamic acid, 2e-3-2,3-dimethoxyphenyl acrylic acid, e-3-2,3-dimethoxyphenyl acrylic acid, 2,3-dimethoxy cinnamic acid, 3-2,3-dimethoxyphenyl acrylic acid, 2e-3-2,3-dimethoxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2,3-dimethoxyphenyl, e-3-2,3-dimethoxyphenyl prop-2-enoic acid, e-3-2,3-dimethoxy-phenyl-acrylic acid
PubChem CID 735842
IUPAC Name (E)-3-(2,3-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=CC(=C1OC)C=CC(=O)O

Specifications

Melting Point 179°C to 183°C
Quantity 250 g
Beilstein 2695122
Formula Weight 208.21
Percent Purity ≥98%
Chemical Name or Material 2,3-Dimethoxycinnamic acid, predominantly trans
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 231-249-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.