Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethoxyphenol, 98%

Catalog No. AA4406004
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4406004 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4406004 Supplier Thermo Scientific Chemicals Supplier No. 04406004
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5150-42-5 | C8H10O3 | 154.165 g/mol

2,3-Dimethoxyphenol is used to study the nitrosative deamination of DNA bases induced by reactive nitrogen species (RNS) while studying the cause of mutagenesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5150-42-5
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00008366
InChI Key QSZCGGBDNYTQHH-UHFFFAOYSA-N
Synonym dimethoxyphenol, phenol, dimethoxy, unii-ow147i6c84, 1-hydroxy-2,3-dimethoxybenzene, pyrogallol 1,2-dimethyl ether, phenol, 2,3-dimethoxy, phenol,3-dimethoxy, 2,3-di-methoxyphenol, 2,3-dim ethoxyphenol, 2,3-dimethoxy phenol
PubChem CID 78828
IUPAC Name 2,3-dimethoxyphenol
SMILES COC1=CC=CC(=C1OC)O

Specifications

Density 1.182 g/mL
Boiling Point 233°C to 234°C
Flash Point 108°C (226°F)
Refractive Index 1.54
Quantity 2 g
Solubility Information Soluble in Chloroform.
Formula Weight 154.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2,3-Dimethoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 225-922-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.