Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-2,3-dinitrobutane 98.0+%, TCI America™
SDP

Catalog No. D182210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D182210G 10 g
D182225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D182210G Supplier TCI America Supplier No. D182210G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3964-18-9
Molecular Formula C6H12N2O4
Molecular Weight (g/mol) 176.172
MDL Number MFCD00007392
InChI Key DWCLXOREGBLXTD-UHFFFAOYSA-N
PubChem CID 77577
IUPAC Name 2,3-dimethyl-2,3-dinitrobutane
SMILES CC(C)(C(C)(C)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 215°C
Color White
Quantity 10 g
UN Number 2926
Formula Weight 176.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethyl-2,3-dinitrobutane
RTECSNumber EJ9943333
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.