Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-dimethyl-2,3-diphenylbutane | 1889-67-4 | MFCD00053713 | 95.0% | 238.37 g/mol | C18H22 | 100 G
SDP

Catalog No. 5000208398
Encompass_Preferred

2,3-Dimethyl-2,3-diphenylbutane is a research reagent and free-radical initiator typically formed as a minor decomposition product of dicumyl peroxide. It is used in studies of radical reactions, polymerization processes, and materials research. For research use only; not for human or clinical use.

  • Purity 95.0%.
  • Molecular weight 238.37 g/mol.
  • Chemical formula C18H22.
  • Cas number 1889-67-4.
  • Used as a free-radical initiator for polymerization and radical-reaction studies.

Catalog No. 50-002-08398 Supplier Medchemexpress LLC Supplier No. HYW087905100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-Dimethyl-2,3-diphenylbutane is a research reagent and free-radical initiator typically formed as a minor decomposition product of dicumyl peroxide. It is used in studies of radical reactions, polymerization processes, and materials research. For research use only; not for human or clinical use.

  • Purity 95.0%.
  • Molecular weight 238.37 g/mol.
  • Chemical formula C18H22.
  • Cas number 1889-67-4.
  • Used as a free-radical initiator for polymerization and radical-reaction studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.