Learn More
Medchemexpress LLC 2,3-dimethyl-2,3-diphenylbutane | 1889-67-4 | MFCD00053713 | 95.0% | 238.37 g/mol | C18H22 | 50 G
SDP
Catalog No. 5000208418
Encompass_Preferred
2,3-dimethyl-2,3-diphenylbutane is a small-molecule research chemical used as a free-radical initiator and a minor decomposition product of dicumyl peroxide. It is applied in polymerization and organic synthesis to promote radical reactions and study decomposition pathways.
  • Acts as a free-radical initiator for polymerization and cross-linking reactions.
  • Commonly formed during thermal decomposition of dicumyl peroxide.
  • Offered at laboratory scales in multiple pack sizes for research use.
  • Typical purity is 95.0% with molecular weight 238.37 g/mol and formula C18H22.
  • Store as a powder at low temperatures to maximize shelf life.
Catalog No. 50-002-08418 Supplier Medchemexpress LLC Supplier No. HYW08790550G
2,3-dimethyl-2,3-diphenylbutane is a small-molecule research chemical used as a free-radical initiator and a minor decomposition product of dicumyl peroxide. It is applied in polymerization and organic synthesis to promote radical reactions and study decomposition pathways.
  • Acts as a free-radical initiator for polymerization and cross-linking reactions.
  • Commonly formed during thermal decomposition of dicumyl peroxide.
  • Offered at laboratory scales in multiple pack sizes for research use.
  • Typical purity is 95.0% with molecular weight 238.37 g/mol and formula C18H22.
  • Store as a powder at low temperatures to maximize shelf life.