Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,3-dimethyl-2,3-diphenylbutane | 1889-67-4 | MFCD00053713 | 95.0% | 238.37 g/mol | C18H22 | 500 G
SDP

Catalog No. 5000208399
Encompass_Preferred

2,3-dimethyl-2,3-diphenylbutane is a hydrocarbon that forms as a minor product during the thermal decomposition of dicumyl peroxide and can act as a free-radical initiator in polymerization and organic synthesis processes.

  • Cas number: 1889-67-4.
  • Purity: 95.0%.
  • Chemical formula: C18H22.
  • Molecular weight: 238.37 g/mol.
  • Physical form: solid powder.
  • Storage: powder at -20°C for up to 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Available package sizes: 50 g, 100 g, 500 g, and bulk options.

Catalog No. 50-002-08399 Supplier Medchemexpress LLC Supplier No. HYW087905500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,3-dimethyl-2,3-diphenylbutane is a hydrocarbon that forms as a minor product during the thermal decomposition of dicumyl peroxide and can act as a free-radical initiator in polymerization and organic synthesis processes.

  • Cas number: 1889-67-4.
  • Purity: 95.0%.
  • Chemical formula: C18H22.
  • Molecular weight: 238.37 g/mol.
  • Physical form: solid powder.
  • Storage: powder at -20°C for up to 3 years or 4°C for 2 years; in solvent at -80°C for 6 months or -20°C for 1 month.
  • Available package sizes: 50 g, 100 g, 500 g, and bulk options.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.