Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethyl-4-fluoronitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D51875G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51875G 5 g
D51871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D51875G Supplier TCI America Supplier No. D51875G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1736-87-4 | C8H8FNO2 | 169.155 g/mol

Chemical Identifiers

CAS 1736-87-4
Molecular Formula C8H8FNO2
Molecular Weight (g/mol) 169.155
MDL Number MFCD05663712
InChI Key GLDMIZKOJPVEIV-UHFFFAOYSA-N
Synonym 2,3-dimethyl-4-fluoronitrobenzene, 3-fluoro-6-nitro-1,2-dimethylbenzene, 3-fluoro-6-nitro-o-xylene, benzene, 1-fluoro-2,3-dimethyl-4-nitro, 3-fluoro-1,2-dimethyl-6-nitrobenzene, 4-fluoro-2,3-dimethyl nitrobenzene, intermediates-zcf02291, ksc534o2t, 2,3-dimethyl-1-fluoro-4-nitrobenzene, 4-fluoro-2,3-dimethyl-1-nitrobenzene
PubChem CID 24721201
IUPAC Name 1-fluoro-2,3-dimethyl-4-nitrobenzene
SMILES CC1=C(C=CC(=C1C)F)[N+](=O)[O-]

Specifications

Melting Point 40°C
Quantity 5 g
Physical Form Crystal-Lump at 20°C
Boiling Point 110°C
Formula Weight 169.16
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2,3-Dimethyl-4-fluoronitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.