Learn More
Aobchem 2 3-Dimethyl-4-(phenylmethoxy)benzaldehyde | ≥97% | CAS 1609289-76-0 | C16H16O2 | 250mg
Catalog No. 5000902447
Encompass_Preferred
2 3-Dimethyl-4-(phenylmethoxy)benzaldehyde | ≥97% | CAS 1609289-76-0 | C16H16O2 | 250mg
Catalog No. 50-009-02447 Supplier Aobchem Supplier No. A33337250MG
2 3-Dimethyl-4-(phenylmethoxy)benzaldehyde | ≥97% | CAS 1609289-76-0 | C16H16O2 | 250mg