Learn More
Sigma Aldrich 2-(3-dimethylaminopropoxy)benzaldehyde
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 2,2-dimethylcyclopentanone is a ketone. synthesis of various c-2 substituted vitamin d derivatives with a 2,2-dimethylcyclopentanone unit in side chains has been reported. regioselective synthesis of 2,2-dimethylcyclopentanone, via its enolate precursor regioselectively obtained using the 2-pyrrolidone magnesium salt, has been reported.2,2-dimethylcyclopentanone enolate may be used as starting reagent in the enantioselective synthesis of chiral phosphines belonging to the p-aryl-2-phosphabicyclo[3.3.0]octane family (pbo). it may be used in the synthesis of:
- 2,6,6-trimethyl-2-azaspiro[4.4]nonane-1,3-dione, a spirosuccinimide moiety of asperparaline a
- Novel spiropentanopyrrolizidine oxime alkaloids, namely 2α,3α,5α,6α,7α,7aα-hexahydro-2,2-dimethylspirocyclopentane-1
- α,α-dimethyl-α-valerolactone, via baeyer-villiger oxidation
Specifications
Specifications
| Molecular Formula | C12H17NO2 |
| Linear Formula | C12H17NO2 |
| Quantity | 5 g |
| MDL Number | MFCD00128460 |
| Molecular Weight (g/mol) | 207.27 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.