Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylaniline Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D067325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D067325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D067325G Supplier TCI America Supplier No. D067325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory reagent grade provided at 98.0% minimum purity.

Chemical Identifiers

CAS 5417-45-8
Molecular Formula C8H12ClN
Molecular Weight (g/mol) 157.641
MDL Number MFCD00054332
InChI Key DUQDEEVOZDZJGQ-UHFFFAOYSA-N
Synonym 2,3-Xylidine Hydrochloride
PubChem CID 22239262
IUPAC Name 2,3-dimethylaniline;hydrochloride
SMILES CC1=C(C(=CC=C1)N)C.Cl

Specifications

Color White
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 157.64
Percent Purity ≥98.0% (HPLC,N)
Grade Not Available
Chemical Name or Material 2,3-Dimethylaniline Hydrochloride
EINECSNumber (3)-0129
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.