Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylanisole, 97%, Thermo Scientific™

Catalog No. AC159995000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC159995000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC159995000 Supplier Thermo Scientific Chemicals Supplier No. 159995000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2944-49-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008376
InChI Key BLMBNEVGYRXFNA-UHFFFAOYSA-N
Synonym 2,3-dimethylanisole, 3-methoxy-o-xylene, benzene, 1-methoxy-2,3-dimethyl, benzene, methoxydimethyl, 1,2-dimethyl-6-methoxybenzene, 1-methoxy-2,3-dimethyl-benzene, dimethylanisole, pubchem4114, 2,3-dimethyl anisole, 2,3-dimethyl-anisole
PubChem CID 76269
IUPAC Name 1-methoxy-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)OC)C

Specifications

Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 0.984g/mL
Quantity 500 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5190 to 1.5220 (20°C, 589nm)
Assay Percent Range 97%
Linear Formula (CH3)2C6H3OCH3
Beilstein 06, 480
Formula Weight 136.19
Percent Purity ≥96%
Chemical Name or Material 2, 3-Dimethylanisole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.