Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylanisole 98.0+%, TCI America™
SDP

Catalog No. D220010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D220010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D220010ML Supplier TCI America Supplier No. D220010ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity 2,3-dimethylanisole with a minimum GC purity of 98.0%.

Chemical Identifiers

CAS 2944-49-2
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00008376
InChI Key BLMBNEVGYRXFNA-UHFFFAOYSA-N
Synonym 2,3-dimethylanisole, 3-methoxy-o-xylene, benzene, 1-methoxy-2,3-dimethyl, benzene, methoxydimethyl, 1,2-dimethyl-6-methoxybenzene, 1-methoxy-2,3-dimethyl-benzene, dimethylanisole, pubchem4114, 2,3-dimethyl anisole, 2,3-dimethyl-anisole
PubChem CID 76269
IUPAC Name 1-methoxy-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)OC)C

Specifications

Color White-Yellow
Quantity 10 mL
Physical Form Crystal-Lump at 20°C
Boiling Point 195°C
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 2,3-Dimethylanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.