Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzeneboronic acid, 98%

Catalog No. AAB2394206
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2394206 5 g
AAB2394203 1 g
AAB2394214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2394206 Supplier Thermo Scientific Chemicals Supplier No. B2394206
Only null left
Add to Cart

CAS: 183158-34-1 | C8H11BO2 | 149.984 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 183158-34-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD01863524
InChI Key ZYYANAWVBDFAHY-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
PubChem CID 2773395
IUPAC Name (2,3-dimethylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)C)C)(O)O

Specifications

Melting Point ∼175°C
Quantity 5 g
Beilstein 8050763
Formula Weight 149.99
Percent Purity 98%
Chemical Name or Material 2,3-Dimethylbenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.