Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylbenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D44965G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D44965G Supplier TCI America Supplier No. D44965G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13651-14-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00046742
InChI Key ZQQIVMXQYUZKIQ-UHFFFAOYSA-N
PubChem CID 96208
IUPAC Name (2,3-dimethylphenyl)methanol
SMILES CC1=CC=CC(CO)=C1C

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 131°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dimethylbenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.