Learn More
Supelco Inc 2,3-Dimethylbutane, 79-29-8, MFCD00008925, 500 mL
Supplier:  Supelco Inc D151602500ML
Encompass
Linear Formula: (CH3)2CHCH(CH3)2, Molecular Weight: 86.18, Synonym: Diisopropyl, Molecular Weight: 86.18, assay: 98%. 2,3-Dimethylbutane can can undergo hydrogenolysis over supported catalysts of ruthenium, nickel, cobalt, and iron to form smaller hydrocarbons, mainly methane.
Catalog No. NC0703405