Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylcyclohexanol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D166025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D166025ML 25 mL
D16605ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D166025ML Supplier TCI America Supplier No. D166025ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1502-24-5
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001444
InChI Key KMVFQKNNDPKWOX-UHFFFAOYSA-N
PubChem CID 102631
IUPAC Name 2,3-dimethylcyclohexan-1-ol
SMILES CC1CCCC(C1C)O

Specifications

Color Colorless
Boiling Point 181°C
Quantity 25 mL
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Dimethylcyclohexanol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.