Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylpentane, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018610 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8610 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8610 Supplier MilliporeSigma Supelco Supplier No. 410855ML
Add to Cart
Add to Cart

2,3-Dimethylpentane belongs to the class of volatile organic compounds (VOCs).

Featured Industries: Environmental
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 639°F
CAS 565-59-3
Vapor Pressure 2.35 psi (37.7°C)
Vapor Density 3.45 (vs. air)
Density 0.695 g/mL (at 25°C)
Flash Point -7°C
Physical Form Neat
Linear Formula C2H5 CH(CH3)CH(CH3)2
MDL Number MFCD00009337
Boiling Point 89°C to 90°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 100.20
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.