Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. D35165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35165G Supplier TCI America Supplier No. D35165G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Organometallic reagent suitable for laboratory research applications.

Chemical Identifiers

CAS 183158-34-1
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.984
MDL Number MFCD01863524
InChI Key ZYYANAWVBDFAHY-UHFFFAOYSA-N
Synonym 2,3-dimethylbenzeneboronic acid, 2,3-dimethylphenyl boronic acid, o-xylene-3-boronic acid, 2,3-dimethylphenyl boranediol, 2,3-dimethylbenzeneboronicacid, 2,3-dimethylphenylboronicacid, pubchem5037, acmc-1bxk2, amtb394, ksc174k9l
PubChem CID 2773395
IUPAC Name (2,3-dimethylphenyl)boronic acid
SMILES B(C1=C(C(=CC=C1)C)C)(O)O

Specifications

Melting Point 175°C
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 149.98
Percent Purity 97.0 to 114.0 %
Grade Not Available
Chemical Name or Material 2,3-Dimethylphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.