Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimethylquinoxaline 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
2379-55-7
C10H10N2
158.2
MFCD00006728
FKHNZQFCDGOQGV-UHFFFAOYSA-N
quinoxaline, 2,3-dimethyl,aurora ka-3449,quinoxaline,3-dimethyl,2,3-dimethyl-quinoxalin,2,3-dimethyl-quinoxaline,acmc-1cg5s,quinoxaline,2,3-dimethyl,salor-int l498777-1ea,5-23-07-00267 beilstein handbook reference,chembl67865
16925
2,3-dimethylquinoxaline
CC1=NC2=CC=CC=C2N=C1C
2379-55-7
C10H10N2
158.2
MFCD00006728
FKHNZQFCDGOQGV-UHFFFAOYSA-N
quinoxaline, 2,3-dimethyl,aurora ka-3449,quinoxaline,3-dimethyl,2,3-dimethyl-quinoxalin,2,3-dimethyl-quinoxaline,acmc-1cg5s,quinoxaline,2,3-dimethyl,salor-int l498777-1ea,5-23-07-00267 beilstein handbook reference,chembl67865
16925
2,3-dimethylquinoxaline
CC1=NC2=CC=CC=C2N=C1C
Infrared Spectrum Authentic
Flash Point >100°C
Packaging Glass bottle
Quantity 25g
Color Beige
Melting Point 102.0°C to 107.0°C
Boiling Point 257.0°C to 259.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 23, 191
Solubility Information Solubility in water: insoluble
Formula Weight 158.2
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2, 3-Dimethylquinoxaline, 97%
Show More Show Less

EINECSNumber : 219-162-0

RTECSNumber : VD1960000

TSCA : TSCA