Learn More
Cayman Chemical 2 3-dInor Thromboxn B2 100ug
SDP
Catalog No. 502982490
Encompass_Preferred
An endogenous CB neurotransmitter that binds to both CB1 and CB2 receptors
Catalog No. 50-298-2490 Supplier Cayman Chemical Supplier No. 19050100 UG
An endogenous CB neurotransmitter that binds to both CB1 and CB2 receptors