Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diphenylpropionic Acid 98.0+%, TCI America™
SDP

Catalog No. D17905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D17905G 5 g
D179025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D17905G Supplier TCI America Supplier No. D17905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3333-15-1
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00044405
InChI Key PCRICPYPVZKEBZ-UHFFFAOYNA-N
PubChem CID 18744
IUPAC Name 2,3-diphenylpropanoic acid
SMILES OC(=O)C(CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 89°C
Color White-Yellow
Boiling Point 340°C
Quantity 5 g
Formula Weight 226.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diphenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.