Learn More
Medchemexpress LLC 2,3-diphospho-D-glyceric acid pentasodium salt | 102783-53-9 | MFCD00070074 | 95.0% | 375.95 g/mol | C3H3Na5O10P2 | 5 MG
SDP
Catalog No. 5000222881
Encompass_Preferred
2,3-Diphospho-D-glyceric acid pentasodium salt is a highly anionic polyphosphorus biochemical reagent (CAS 102783-53-9) used in research to study red blood cell physiology and metabolic pathways. It binds hemoglobin and reduces its oxygen affinity, and is employed in biochemical and cellular assays.
Catalog No. 50-002-22881 Supplier Medchemexpress LLC Supplier No. HY1388225MG
2,3-Diphospho-D-glyceric acid pentasodium salt is a highly anionic polyphosphorus biochemical reagent (CAS 102783-53-9) used in research to study red blood cell physiology and metabolic pathways. It binds hemoglobin and reduces its oxygen affinity, and is employed in biochemical and cellular assays.