Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Epoxy-1-(1-ethoxyethoxy)propane

Catalog No. SYX002751G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX002751G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX002751G Supplier Sigma Aldrich Supplier No. SYX002751G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Kinetic and thermodynamic analysis of catalytic hydrodechlorination of 1-chlorooctadecane in supercritical carbon dioxide using 5% pd supported on α-al2o3 has been reported
  • Incorporation of 1-chlorooctadecane into a host monolayer of stearic acid has been reported.1-chlorooctadecane has been used as internal standard for the determination of endocrine disrupters in water samples by stir bar sorptive extraction method

Specifications

CAS 4416-85-7
Molecular Formula C7H14O3
Linear Formula C7H14O3
MDL Number MFCD22575236
Quantity 25 g
Molecular Weight (g/mol) 146.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.