Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2-[(3-hydroxy-4-nitrophenyl)methylidene]propanedinitrile | 118409-62-4 | MFCD00236448 | 98.0% | 215.17 g·mol^-1 | C10H5N3O3 | 1 ML
SDP

Catalog No. 5000244302
Encompass_Preferred

AG126 (Tyrphostin AG126) is a small-molecule tyrosine kinase inhibitor that inhibits phosphorylation of ERK1 and ERK2 at approximately 25-50 μM. It is supplied for research use in solid form and as DMSO solutions, and is used in studies of meiosis, mitosis, and postmitotic cellular processes.

  • High purity: 98.0%.
  • Available as solid and as 10 mM solution in DMSO.
  • Chemical formula C10H5N3O3; molecular weight 215.17 g·mol⁻¹.
  • Appearance: light yellow to yellow solid.
  • Storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 2 years).

Catalog No. 50-002-44302 Supplier Medchemexpress LLC Supplier No. HY10833010MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

AG126 (Tyrphostin AG126) is a small-molecule tyrosine kinase inhibitor that inhibits phosphorylation of ERK1 and ERK2 at approximately 25-50 μM. It is supplied for research use in solid form and as DMSO solutions, and is used in studies of meiosis, mitosis, and postmitotic cellular processes.

  • High purity: 98.0%.
  • Available as solid and as 10 mM solution in DMSO.
  • Chemical formula C10H5N3O3; molecular weight 215.17 g·mol⁻¹.
  • Appearance: light yellow to yellow solid.
  • Storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solvent -80 °C (up to 2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.