Learn More
Cayman Chemical 2,3-Indolobetulonic Acid, Purity: ≥95%, CAS: 905837-93-6, 10 mg
SDP
Catalog No. 502946587
Encompass_Preferred
An inhibitor of α-glucosidase (IC50 = 1.8 µM)
Catalog No. 50-294-6587 Supplier Cayman Chemical Supplier No. 3306510 MG
An inhibitor of α-glucosidase (IC50 = 1.8 µM)