Learn More
Cayman Chemical 2,3-Isopropylidene-D-ribonolactone, Purity: ≥95%, CAS: 30725-00-9, 1 g
SDP
Catalog No. 502992628
Encompass_Preferred
A building block; has been used in the stereocontrolled synthesis of 6-epi-trehazolin and 6-epi-trehalamine
Catalog No. 50-299-2628 Supplier Cayman Chemical Supplier No. 304791 G
A building block; has been used in the stereocontrolled synthesis of 6-epi-trehazolin and 6-epi-trehalamine