Learn More
Medchemexpress LLC 2',3'-Isopropylideneguanosine | 362-76-5 | 323.31 | 25 G
SDP
Catalog No. 5000448179
Encompass_Preferred
2',3'-Isopropylideneguanosine is a biochemical assay reagent that acts as a precursor for m7,20,30-isopropylideneG[5']ppp[5']G.
  • Functions as a biochemical assay reagent
  • Precursor in the synthesis of m7,20,30-isopropylideneG[5']ppp[5']G
Catalog No. 50-004-48179 Supplier Medchemexpress LLC Supplier No. 25GHYW008240
2',3'-Isopropylideneguanosine is a biochemical assay reagent that acts as a precursor for m7,20,30-isopropylideneG[5']ppp[5']G.
  • Functions as a biochemical assay reagent
  • Precursor in the synthesis of m7,20,30-isopropylideneG[5']ppp[5']G