Learn More
Cayman Chemical 2,3-MDA (hydrochloride), Purity: ≥98%, CAS: 86029-48-3, 50 mg
SDP
Catalog No. 502954258
Encompass_Preferred
A structural homolog of MDA which differs only in the placement of the methyledioxy group; intended for forensic and research applications
Catalog No. 50-295-4258 Supplier Cayman Chemical Supplier No. 1396950 MG
A structural homolog of MDA which differs only in the placement of the methyledioxy group; intended for forensic and research applications