Learn More
Cayman Chemical 2 3-MDMAhydrochlorIde 10mg
SDP
Catalog No. 502954232
Encompass_Preferred
An aromatic positional isomer of 3,4-MDMA (ecstasy) that exhibits a similar potency to 3,4-MDMA for inhibition of the noradrenalin transporter (IC50s = 6.2 and 6.6 μM, respectively) but is less potent than 3,4-MDMA for inhibition of the serotonin transporter (IC50s = 82 μM and 34.8 μM, respectively)
Catalog No. 50-295-4232 Supplier Cayman Chemical Supplier No. 1397010 MG
An aromatic positional isomer of 3,4-MDMA (ecstasy) that exhibits a similar potency to 3,4-MDMA for inhibition of the noradrenalin transporter (IC50s = 6.2 and 6.6 μM, respectively) but is less potent than 3,4-MDMA for inhibition of the serotonin transporter (IC50s = 82 μM and 34.8 μM, respectively)