Learn More
Medchemexpress LLC 2-(3-mercaptopropyl)pentanedioic acid | 254737-29-6 | MFCD24479659 | 98.0% | 206.26 g/mol | C8H14O4S | 5 MG
SDP
Catalog No. 5000244917
Encompass_Preferred
2-(3-mercaptopropyl)pentanedioic acid is an orally active, selective inhibitor of glutamate carboxypeptidase II (GCP II/NAALADase) used in biochemical and in vivo pharmacology research. It is supplied as a small-molecule standard for laboratory studies and is characterized by a thiol-containing structure that confers oral bioavailability.
Catalog No. 50-002-44917 Supplier Medchemexpress LLC Supplier No. HY1033455MG
2-(3-mercaptopropyl)pentanedioic acid is an orally active, selective inhibitor of glutamate carboxypeptidase II (GCP II/NAALADase) used in biochemical and in vivo pharmacology research. It is supplied as a small-molecule standard for laboratory studies and is characterized by a thiol-containing structure that confers oral bioavailability.