Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Methoxyphenethyl)phenol 98.0+%, TCI America™
SDP

Catalog No. M24725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24725G 5 g
M24721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24725G Supplier TCI America Supplier No. M24725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 167145-13-3
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.291
MDL Number MFCD09800498
InChI Key HGQQRAXOBYWKDV-UHFFFAOYSA-N
Synonym 2-[2-(3-Methoxyphenyl)ethyl]phenol
PubChem CID 10585448
IUPAC Name 2-[2-(3-methoxyphenyl)ethyl]phenol
SMILES COC1=CC=CC(=C1)CCC2=CC=CC=C2O

Specifications

Melting Point 47°C
Color White
Quantity 5 g
Formula Weight 228.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Methoxyphenethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.