Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Methylphenyl)benzimidazole, 95%, Thermo Scientific™

Catalog No. AAH64264MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64264MD 250 mg
AAH6426403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64264MD Supplier Thermo Scientific Chemicals Supplier No. H64264MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6528-83-2
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD01220491
InChI Key USLJYASOSQJYGN-UHFFFAOYSA-N
Synonym 2-3-methylphenyl benzimidazole, 2-3-methylphenyl-1h-benzimidazole, 2-m-tolyl-1h-benzo d imidazole, 2-3-methylphenyl-1h-1,3-benzodiazole, 2-3-methylphenyl-1h-benzo d imidazole, 2-m-tolyl-1h-benzoimidazole, cbmicro_005935, cambridge id 5978943, 2-3-methylphenyl-benzimidazole
PubChem CID 2879476
IUPAC Name 2-(3-methylphenyl)-1H-benzimidazole
SMILES CC1=CC=CC(=C1)C2=NC3=CC=CC=C3N2

Specifications

Color White
Quantity 250 mg
Formula Weight 208.26
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(3-Methylphenyl)benzimidazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.