Learn More
BioSpec Products 2.3 MM SS BEADS, 90 GRAM PACK
SDP
Catalog No. NC0792563
Encompass_Preferred
Stainless Steel Beads, 2.3mm dia 90g Stainless steel beads have a density of 7.9g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available. *Note: These beads are not sold in one pound amounts.
Catalog No. NC0792563 Supplier BioSpec Products Supplier No. 11079123SS
Stainless Steel Beads, 2.3mm dia 90g Stainless steel beads have a density of 7.9g/cc. They are used mostly for grinding leaves and seeds. They are made of Type 316 stainless steel - the most corrosion resistant available. *Note: These beads are not sold in one pound amounts.