Learn More
Medchemexpress LLC 2,3-Naphthalenediamine | 771-97-1 | 98.0% | 1 G
SDP
Supplier:  Medchemexpress LLC HYD00731G
Encompass_Preferred
2,3-Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium ion detection and for the fluorometric determination of nitrite. It is for research use only. Poly(2,3-diaminonaphthalene) microspheres have been utilized as a novel quencher for fluorescence-enhanced nucleic acid detection.
  • Highly selective colorimetric and fluorometric reagent for selenium ion detection.
  • Used for the fluorometric determination of nitrite.
  • Acts as a quencher for fluorescence-enhanced nucleic acid detection when in microsphere form.
  • Used in kinase assays to quantify N-nitrosation.
Catalog No. 50-004-10813