Learn More
Medchemexpress LLC 2,3-naphthalenedicarboxaldehyde | 7149-49-7 | MFCD00016618 | 99.3% | 184.19 g/mol | C12H8O2 | 1 G
SDP
Catalog No. 5000302155
Encompass_Preferred
Naphthalene-2,3-dicarboxaldehyde is an aromatic dialdehyde used as a research reagent and derivatization agent. It reacts with primary amines to form highly fluorescent products and is applied in analytical workflows for detection and quantification, as well as in colorimetric assays.
  • Forms fluorescent derivatives with primary amines for sensitive detection.
  • Compatible with chromatographic and spectrofluorimetric analysis methods.
  • Used in colorimetric determination of cholesterol and other assays.
  • High-purity grades available for analytical applications.
  • Solid reagent; store and handle according to safety data sheet recommendations.
Catalog No. 50-003-02155 Supplier Medchemexpress LLC Supplier No. HYW0162881G
Naphthalene-2,3-dicarboxaldehyde is an aromatic dialdehyde used as a research reagent and derivatization agent. It reacts with primary amines to form highly fluorescent products and is applied in analytical workflows for detection and quantification, as well as in colorimetric assays.
  • Forms fluorescent derivatives with primary amines for sensitive detection.
  • Compatible with chromatographic and spectrofluorimetric analysis methods.
  • Used in colorimetric determination of cholesterol and other assays.
  • High-purity grades available for analytical applications.
  • Solid reagent; store and handle according to safety data sheet recommendations.