Learn More
Cayman Chemical 2 3-O- 5-BromonthrnIloyl 3.5mg
SDP
Supplier:  Cayman Chemical 384453.5 MG
Encompass_Preferred
Fluorescent ATP derivatives; inhibits mammalian AC1, AC2, and AC5 and B. pertussis AC toxin CyaA (Kis = 120, 450, 70, and 330 nM, respectively)
Catalog No. 50-291-8249