Learn More
Cayman Chemical 2'(3')-O-(5-Bromoanthraniloyl)inosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.5 mg
SDP
Catalog No. 502917113
Encompass_Preferred
Fluorescent ITP derivatives; selectively inhibits mammalian AC1, AC2, and AC5 over B. pertussis CyaA (Kis = 7, 22, 4.6, and 920 nM, respectively)
Catalog No. 50-291-7113 Supplier Cayman Chemical Supplier No. 384463.5 MG
Fluorescent ITP derivatives; selectively inhibits mammalian AC1, AC2, and AC5 over B. pertussis CyaA (Kis = 7, 22, 4.6, and 920 nM, respectively)