Learn More
Cayman Chemical 2'(3')-O-(5-Chloroanthraniloyl)inosine-5'-O-triphosphate (sodium salt), Purity: ≥95% (mixture of isomers), 3.3 mg
SDP
Catalog No. 502918250
Encompass_Preferred
Fluorescent ITP derivatives; selectively inhibits mammalian AC1, AC2, and AC5 over B. pertussis AC toxin CyaA (Kis = 3.3, 8, 2, and 700 nM, respectively)
Catalog No. 50-291-8250 Supplier Cayman Chemical Supplier No. 384503.3 MG
Fluorescent ITP derivatives; selectively inhibits mammalian AC1, AC2, and AC5 over B. pertussis AC toxin CyaA (Kis = 3.3, 8, 2, and 700 nM, respectively)